Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
98.00 DKK - 481.00 DKK
Product Code | Brand | Capacity (Metric) | Closure Size | Tolerance | Neck Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Tolerance | Neck Style | Price | Quantity & Availability | ||
10033861
|
Karl Hecht™
42096076 |
500 mL | NS 19/26 | 0.25 mL | Standard |
202.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10648671
|
Karl Hecht™
42096052 |
100 mL | NS 14/23 | 0.10 mL | Standard |
125.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10195331
|
Karl Hecht™
42096004 |
5 mL | NS 7/16 | 0.025 mL | Standard |
126.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10776372
|
Karl Hecht™
42096016 |
10 mL | NS 10/19 | 0.4 mL | Wide |
125.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10339021
|
Karl Hecht™
42096024 |
20 mL | NS 12/21 | 0.06 mL | Wide |
138.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10490172
|
Karl Hecht™
42096028 |
25 mL | NS 10/19 | 0.04 mL | Standard |
138.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10459421
|
Karl Hecht™
42096060 |
200 mL | NS 14/23 | 0.15 mL | Standard |
163.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10748682
|
Karl Hecht™
42096064 |
250 mL | NS 14/23 | 0.15 mL | Standard |
171.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10359211
|
Karl Hecht™
42096084 |
1000 mL | NS 29/32 | 0.6 mL | Standard |
389.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10013791
|
Karl Hecht™
42096040 |
50 mL | NS 12/21 | 0.06 mL | Standard |
98.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10083871
|
Karl Hecht™
42110018 |
5 mL | NS 7/16 | 0.025 mL |
149.00 DKK
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
10145481
|
Karl Hecht™
42096012 |
10 mL | NS 7/16 | 0.025 mL | Standard |
125.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10349261
|
Karl Hecht™
42110012 |
2 mL | NS 7/16 | 0.025 mL |
134.00 DKK
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
10708852
|
Karl Hecht™
42096088 |
2000 mL | NS 29/32 | 0.6 mL | Standard |
481.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10638671
|
Karl Hecht™
42096044 |
50 mL | NS 14/23 | 0.10 mL | Wide |
98.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Duran™ borosilicate glass 3.3.
- Standard type.
- Standard ground and interchangeable NS stopper of polyethylene.
- Fused blue graduation.
- Thick-walled, solid finish.
- Clearly legible figures and marks.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Polyethylene | |
NS Stopper |