Learn More
Buerkle™ Wall-mount Draining Rack
Designed with individually placed stable polypropylene rods. Buerkle™ Wallmount Draining Racks can be configured to fit larger or smaller apparatus.
Brand:  Buerkle™ 9640-2050
 View more versions of this product
Product Code. 10662921
  • 2435.00 DKK / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Description
A wall-mounted draining rack, including a wall mounting set (screws, dowels). A removable draining channel is optionally available, with a connection for a draining tube which can discharge directly into a laboratory wash basin.
  • Draining rack PVC, rods polypropylene
  • Including spanner for the rods
  • Three different rod lengths to adapt to varying apparatus
  • Shatterproof
  • Rods click into place for secure positioning
Specifications
Specifications
Spanner, wall mounting set of screws and dowels, removable draining channel, connection for a draining tube, box for the rods, 10 x 60 mm rods, 39 x 100 mm rods, 4 x 150 mm rods
PVC Drain Rack, Polypropylene Rods
500 mm
500 mm
With draining channel
77
19.68 in.
19.68 in.
Videos
  • Buerkle Wall Mounted Draining Rack
    Buerkle Wall Mounted Draining Rack
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle Wall Mounted Draining Rack
    Certificates
    View accessory products
    Product Title
    Buerkle™ Wall-mount Draining Rack > Size: 500 x 500 mm; With draining channel