missing translation for 'onlineSavingsMsg'
Learn More
Water, HPLC Gradient Grade, Scharlab Product Code.: 11362625
Estimated Shipment: 12-06-2025
Add to basket

Water, HPLC Gradient Grade, Scharlab

Product Code. 11362625
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11362625

Brand: Scharlab AG00011000

Estimated Shipment: 12-06-2025
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for use in solvents and in analytical chemistry, including HPLC.

  • Molecular Formula: H2O
  • Molar Mass: 18.02g/mol
  • CAS: 7732-18-5
  • EIN ECS No.: 231-791-2
  • Microfiltered through membranes of pore diameter 0.22μm
  • Colony Count: max. 25 UFC/g
  • Microbiological Test: Passes test

Specifications

CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity 1μS/cm max at 25°C
Molecular Formula H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02g/mol
Grade HPLC Gradient
Residue after Evaporation 0.0001% max
Vapor Pressure 23hPa (at 20°C)
Density 1.00g/cm3
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, HPLC Gradient Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.