Learn More
Usbeck Laborgeraete™ Water Jet Pump
Brand:  Usbeck Laborgeraete™ 5700
 View more versions of this product
Product Code. 10515682
  • 434.07 DKK / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Specifications
145 g
Product Title
Usbeck Laborgeraete™ Water Jet Pump > Weight: 145g