Learn More
Usbeck™ Water Jet Pump with Female Connection to Water Tap Diameter
Impact your filtration process. Usbeck™ Water Jet Pump with Female Connection to Water Tap Diameter are designed to help regulate and control filtration.
Brand:  Usbeck™ 5722
Product Code. 10659531
  • 572.05 DKK / Each
Only null left
Estimated Shipment: 15-09-2025
Description
  • Nickel plated brass
  • Non-return valve
  • Female Connection to
    water tap Ø outside
Specifications
Specifications
26.5 mm
Nickel-plated Brass
Female Connection
250 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck™ Water Jet Pump with Female Connection to Water Tap Diameter >