Learn More
Jaytec Glass™ Weighing Funnel for Cream or Cheese According to BS.696
The Weighing Funnel for Cream or Cheese According to BS.696 is used for weighing out cream or cheese for Gerber tests. Complete with Stopper.
Brand:  Jaytec Glass™ MP.710
Product Code. 12304016
  • 384.00 DKK / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Description
  • Borosilicate glass
  • 7/16 joint on stopper
Specifications
Specifications
19 mL
Funnel for weighing cream or cheese
Borosilicate Glass
75 mm