Learn More
Eppendorf™ Weighing Vessel
Brand: Eppendorf™ 3113807007
230.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Weighing Vessel | |
For Use With Calibration Kit | |
24 mm |
Product Content Correction
Product Title
Eppendorf™ Weighing Vessel > Diameter: 24mm
Spot an opportunity for improvement?Share a Content Correction