Learn More
Tocris Bioscience™ WH-4-023
Potent and selective Lck and Src inhibitor; also inhibits SIK
Brand: Tocris Bioscience™ 5413/50
This item is not returnable.
View return policy
Description
WH-4-023 is a potent and selective Lck and Src inhibitor (IC50 values are 2 and 6 nM respectively). Exhibits >300-fold selectivity against p38α and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. Supports self-renewal of naive hESCs in combination with PD 0325901, CHIR 99021 and SB 590885 .
Specifications
>98% | |
50 mg | |
568.67 |
KIN112 | |
WH-4-023 | |
WH-4-023 |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction