Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
Brand: Cytiva 10417139
6935.00 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low affinity for stains improves visibility under microscope
- Low protein binding minimizes sample loss of proteinaceous samples
- Narrow pore size distribution and high pore density support rapid surface capture of analyte
- Negligible absorption and adsorption of filtrate; nonhygroscopic
- No particle shedding enables clean filtrate
- Biologically inert
Specifications
Polycarbonate | |
Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology | |
Membrane Circle |
0.4 μm | |
25 Pk. | |
293 mm |