Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Featuring sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters
Brand:  Cytiva 10417139
6935.00 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17169880
  • 7300.00 DKK / Pack of 25
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Description
  • Low affinity for stains improves visibility under microscope
  • Low protein binding minimizes sample loss of proteinaceous samples
  • Narrow pore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate
Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology
Membrane Circle
0.4 μm
25 Pk.
293 mm