Learn More
Cytiva Whatman™ Quartz Microfiber Tapered Thimble
28 x 70 mm tapered thimble
Brand:  Cytiva 2812-287
1111.50 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 17109890
  • 1170.00 DKK / Pack of 10
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Description
  • 28 x 70 mm thimble
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 Pk.
Quartz
Pollution monitoring techniques
Microfiber Thimble