Learn More
Abnova™ Wheat Germ agglutinin native Protein from wheat germ
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
Formulation | Lyophilized from 0.05M Tris-HCl, 0.15M NaCl, pH 8.6 |
Quantity | 5 mg |
Regulatory Status | RUO |
Content And Storage | Store at -20°C. After reconstitution with deionized water containing 0.9% sodium chloride, store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Form | Lyophilized |
For Use With (Application) | Enzyme Activity, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis |
Purity or Quality Grade | ≥94% |
Host Species | Plants |
Product Type | Agglutinin (Wheat Germ) Native Protein |
Product Content Correction
Product Title
Abnova™ Wheat Germ agglutinin native Protein from wheat germ >
Spot an opportunity for improvement?Share a Content Correction