Learn More
Novus Biologicals™ Wheat Germ Agglutinin (WGA), Fluorescein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
Antigen | Wheat Germ Agglutinin (WGA), Fluorescein |
Concentration | 5 mg/mL |
Quantity | 10 mg |
Content And Storage | Store at 4°C. |
Buffer | 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM CaCl2 |
Product Content Correction
Product Title
Novus Biologicals™ Wheat Germ Agglutinin (WGA), Fluorescein >
Spot an opportunity for improvement?Share a Content Correction