Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand: DWK Life Sciences 224191
1025.99 DKK valid until 2025-12-31
Use promo code "24362" to get your promotional price.
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Natural | |
1000/Cs. | |
20 mm |
Aluminum | |
Solid Top Cap |