Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
851.00 DKK - 1375.00 DKK
Product Code | Brand | Color | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Closure Size | Price | Quantity & Availability | |||||
10194172
![]() |
DWK Life Sciences
224183-01 |
Natural | 20 mm |
851.00 DKK
Pack of 1000 |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
10022372
![]() |
DWK Life Sciences
224182-01 |
Natural | 13 mm |
931.00 DKK
Pack of 1000 |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
10331772
|
DWK Life Sciences
224187-01 |
Natural | 30 mm |
1375.00 DKK
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
11788779
![]() |
DWK Life Sciences
224182-07 |
Green | 13 mm |
1210.00 DKK
Pack of 1000 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10532444
![]() |
DWK Life Sciences
224183-07 |
Green | 20 mm |
1165.00 DKK
Pack of 1000 |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Aluminum | |
1000/Cs. | |
Center Disc Tear-out Cap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined