Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components
Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)
440.00 DKK - 1450.00 DKK
Specifications
For Use With (Equipment) Cyanide distillation kit
View More Specs
Products 5
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
12481551
View Documents
DWK Life Sciences
377164
Cold Finger Jacket
1250.00 DKK
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
12471551
View Documents
DWK Life Sciences
377163
Cold Finger Condenser
995.00 DKK
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
12491551
View Documents
DWK Life Sciences
377165
Distilling Flask
1135.00 DKK
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
12461551
View Documents
DWK Life Sciences
377162
Absorber Top
1450.00 DKK
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
12401561
View Documents
DWK Life Sciences
377166
Inlet Tube
440.00 DKK
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
Description
Description
Compliance
For use according to ASTM D 2036
Specifications
Specifications
Cyanide distillation kit
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components