Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Kit
Removes interference substances in cyanide quantitative analyses according to ASTM™ D 2036
Brand:  DWK Life Sciences 377160
4739.67 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11792763
  • 5170.00 DKK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
Estimated Shipment: 01-04-2026
Add to basket
Includes: Absorber jacket, absorber top, cold finger condenser, cold finger jacket, 1000mL distilling flask, and inlet tube
Description
Description
Sample reacts with HS2O4 and MgCl2 to convert any cyanide to hydrocyanic acid (HCN). HCN is distilled and absorbed in NaOH solution, and sample is analyzed for CN.
  • With standard taper ClearSeal™ grease-free joints and medium frit
  • Wheaton-33™ low extractable borosilicate glass
  • Components also available separately; see cat. no. 091081
Compliance
For use according to ASTM D 2036
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP Type I, ASTM E438 Type I, Class A
Absorber jacket, absorber top, cold finger condenser, cold finger jacket, 1000mL distilling flask, and inlet tube, ClearSeal™ grease-free joints and medium frit
Cyanide distillation kit
Ideal for removing interfering substances in quantitative analysis for cyanide
Borosilicate Glass
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Kit > Cyanide distillation kit