Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand:  DWK Life Sciences W988004-S
 View more versions of this product
Product Code. 17902581
  • 6745.00 DKK / Pack of 285
Only null left
Estimated Shipment: 21-08-2025
Description
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
Ultra Pure Straight Plug
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
285/Cs.
Wheaton CompletePAK
13 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers > 13 mm Ultra Pure Serum Stopper, 285/Cs.