Learn More
DWK Life Sciences Wheaton™ E-Z Seals™ Aluminum Vial Caps
Lined seals for use with crimp-top vials
Brand: DWK Life Sciences 224219-05
This item is not returnable.
View return policy
Description
- Ideal closures for chromatography and other instrumentation applications
- Open top seal provides for filling or retrieving contents with a syringe
Specifications
PTFE/Silicone | |
Aluminum | |
1000 Cs. | |
11 mm |
Blue | |
E-Z Seal | |
Open Top Cap |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ E-Z Seals™ Aluminum Vial Caps > Aluminum Seal; For Mouth OD: 11mm; With PTFE/ silicone liner
Spot an opportunity for improvement?Share a Content Correction