Learn More
DWK Life Sciences Wheaton™ Peristaltic Pump Tubing: Sinkers
For use with Wheaton Omnispense™ Elite and Unispense™ PRO peristaltic pumps
Brand: DWK Life Sciences 374320
1505.75 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Available in small and large sizes.
Specifications
Glass | |
Dispensing pump | |
Small Sinker |
Dispensing pump | |
Small |
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1425413 | Certificate of Compliance | 26/03/2025 | 374320 |
1425068 | Certificate of Compliance | 25/03/2025 | 374320 |
1422847 | Certificate of Compliance | 18/02/2025 | 374320 |
1419947 | Certificate of Compliance | 08/01/2025 | 374320 |
1417069 | Certificate of Compliance | 20/12/2024 | 374320 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
1Minutes
53Seconds
29
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Peristaltic Pump Tubing: Sinkers > Sinker; Small; 2 and 3mm
Spot an opportunity for improvement?Share a Content Correction