Learn More
DWK Life Sciences Wheaton™ Reaction Flask With 24/40 Joint
Silver diethyldithiocarbamate method
Brand: DWK Life Sciences 635965
1532.26 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Freestanding
- Wheaton 33 low extractable borosilicate glass
- 200mL
- 24/40 joint
Specifications
200 mL | |
Borosilicate Glass | |
Reaction flask | |
24/40 |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Reaction Flask With 24/40 Joint > 200mL; 24/40 joint
Spot an opportunity for improvement?Share a Content Correction