Learn More
DWK Life Sciences Wheaton™ Snap Plug Caps
Manufactured from Polyethylene
Brand: DWK Life Sciences 242815
983.25 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Closure Style: Snap Cap
- Liner Material: TFE
Specifications
Polypropylene | |
Natural | |
LDPE | |
15 mm |