Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Tear-Off Seals
Tear-off seal can be removed completely allowing content to pour from bottle
Brand: DWK Life Sciences 224192-01
935.68 DKK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Tear-off seal can be removed completely allowing content to pour from bottle
- Aluminum
- Unlined
Specifications
Natural | |
1000/Cs. | |
13 mm |
Aluminum | |
Tear-off Cap |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24