Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Tear-Off Seals
Tear-off seal can be removed completely allowing content to pour from bottle
Brand: DWK Life Sciences 224193-07
This item is not returnable.
View return policy
Description
Tear-off seal can be removed completely allowing content to pour from bottle
- Aluminum
- Unlined
Specifications
Green | |
1000/Cs. | |
20 mm |
Aluminum | |
Complete Tear-off Cap |
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Unlined Aluminum Tear-Off Seals > 20mm (0.79in.); Green
Spot an opportunity for improvement?Share a Content Correction