missing translation for 'onlineSavingsMsg'
Learn More

Grathwol White Bucket, 11.4 L

Used for storing purposes.

Brand:  Grathwol 321575040

Encompass

Product Code. 12397637

  • 34.47 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used for storing purposes
  • Lid not included; sold separately (Cat. No. 12367657)
Specifications

Specifications

Bucket
Lid dia: 295 mm
Bucket only. (Lid is sold separately, see 12367657.)
White
242 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers