Learn More
Thermo Scientific™ White Disposable Shaker/Incubator Trays
Support the Thermo Scientific™ Oxoid™ Signal™ Blood Culture System during shaking and incubation using these disposable shaker/incubator trays.
Brand: Thermo Scientific™ BC0104A
354.55 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Support the Thermo Scientific™ Oxoid™ Signal™ Blood Culture System during shaking and incubation using Thermo Scientific™ Oxoid™ Signal™ Disposable Shaker/Incubator Trays. The Signal Blood Culture System is a unique one bottle manual media system designed to detect aerobic, anaerobic and microaerophilic organisms.
Signal Blood Culture System (BC0100M) comes complete with blood culture bottles filled with the blood culture medium and growth indicator devices. Incubator Trays can be ordered separately, where required.
A single white Disposable Shaker/ Incubator Tray can hold up to 10 bottles.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Disposable Shaker/Incubator Trays | |
5/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6162002 | Certificate of Analysis | 16/08/2024 | BC0104A |
6155145 | Certificate of Analysis | 26/07/2024 | BC0104A |
6107582 | Certificate of Analysis | 18/04/2024 | BC0104A |
3793390 | Certificate of Analysis | 10/02/2024 | BC0104A |
3618221 | Certificate of Analysis | 01/02/2023 | BC0104A |