missing translation for 'onlineSavingsMsg'
Learn More

White Standard Buffer Solutions pH 12.47, WTW™

PTB/NIST traceable standard buffer solutions (work reference buffer solutions) for precision applications that is safe use with a high accuracy of ±0.02 pH

Brand:  WTW 109400

251.43 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10077800

  • 259.00 DKK / 250mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • PTB/NIST traceable standard buffer solutions (work reference buffer solutions) for precision applications - safe use
  • Standard buffering solutions as per DIN 19266-01: 2000
  • Universal buffering solutions
  • High accuracy: ±0.02 pH
  • For precision applications

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100°C
DIN / NIST
Precision applications
Liquid
Bottle
White
1g/cm3
12.47
250 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
White Standard Buffer Solutions pH 12.47, WTW™ > Quantity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.