Learn More
Uvex™ White Type 5/6 Air Disposable Chemical Protection Coveralls
Disposable coveralls constructed of very light, air-permeable and highly breathable SMS material, making them ideal for warm work environments.
Brand:  Uvex™ 9817310
2130.06 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17717719
  • 2660.00 DKK / Pack of 50
Only null left
Estimated Shipment: 30-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Exceptional wearer comfort guaranteed by breathable and lightweight SMS material
  • Particle-tight and spray-tight to a limited extent
  • Ideal for warm working environments and extended periods of wear
  • Free of silicone and paint wetting impairment substances
  • Material: Polypropylene SMS
  • Middle finger loops prevent sleeves sliding up the arm
  • Optimal protection in dusty environments
  • Self-adhesive zipper flap for optimal protection
  • Anti-static properties
  • Excellent moisture management
  • Optimal fit with elasticated waistband
  • Three-part hood for a good fit and unrestricted vision
  • Secure and convenient closures thanks to elasticated bands on hood, arms and legs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Elastic
Covered Elastic
Standard
Taped
Chemicals
Controlled Environments
Medium
Polypropylene
Single-use
Open
Hooded
Zipper with Self-adhesive Flap
Non-sterile
ISO 13982-1 - type 5, EN 13034 - type 6, EN 1149-5, EN 1073-2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Uvex™ White Type 5/6 Air Disposable Chemical Protection Coveralls > Size: Medium