Learn More
BRAND™ Wide Neck Glass Erlenmeyer Flasks
With beaded rim and graduations
Brand: BRAND™ 92844
1048.25 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from Boro 3.3 glass
- Wide neck for ease of filling
- Acc. to DIN EN ISO 24450
- Provided in packs of 10
Specifications
50 mm | |
Borosilicate Glass (3.3) | |
10 |
500 mL | |
175 mm |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now
Product Content Correction
Product Title
BRAND™ Wide Neck Glass Erlenmeyer Flasks > Capacity (Metric): 500 mL
Spot an opportunity for improvement?Share a Content Correction