Learn More
Kautex™ Wide Neck PVC Containers, Transparent with Color
View your bottle contents through the tinted dark color of Kautex™ Wide Neck PVC Containers, Transparent with Color. The square PVC bottle is synthetic.
Brand: Kautex™ 2000070698
This item is not returnable.
View return policy
Description
- Square
- Brown or Amber
- PVC
- Without screw closure
Specifications
100 mL | |
Square | |
Packaging, Dispensing, Sending Samples, Storage, Mailing | |
Yes | |
No | |
71 mm |
Brown | |
31 mm | |
PVC | |
Square | |
Square | |
Wide Neck Container |