Learn More
Brady™ Wire and Cable Marking Labels
Made from nylon cloth.
Brand:  Brady™ M71-30-499
Product Code. 11755663
Description
Description
  • White color
Specifications
Specifications
BMP71 Label Printer Labels
Nylon
19.05 x 38.1 mm
0
38.10 mm
White
1
22.23 mm
19.05 x 38.10 mm
19.05 mm
Documents
One moment while we fetch your results.
Product Title
Brady™ Wire and Cable Marking Labels >