Learn More
Fisherbrand™ Wire Gauze
Provides excellent support to beakers and other glassware during heating on ring stands.Fisherbrand™ Wire Gauze is a flat square manufactured from galvanized iron mesh.
Brand:  Fisherbrand™ 9072
Product Code. 11789806
  • 227.00 DKK / Pack of 10
Only null left
Estimated Shipment: 18-09-2025
Add to basket
Description
Description
  • Non-fraying edges
  • Durable and long-lasting
Specifications
Specifications
Wire Gauze
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Wire Gauze > Size: 125 x 125mm