Learn More
Endecotts™ Woven Wire Mesh Sieve
Available in a variety of diameters and apertures
Brand:  Endecotts™ 200SIW.600
 View more versions of this product
Product Code. 11557484
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Endecotts Woven Wire Mesh Sieves are made of stainless steel and available in a wide variety of diameters and apertures.
  • Woven Wire Mesh Sieve
  • Made of Stainless Steel
Specifications
Specifications
600 μm
7.87 in.
200 mm
30 Mesh
30
Sieve
Stainless Steel
Stainless Steel
Woven Mesh
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Endecotts™ Woven Wire Mesh Sieve > Diameter: 200 mm, Pore Size: 600 μm