Learn More
ABEBA™ X-Light Black Occupational Shoes O2 ESD
Black X-light occupational shoes with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Brand:  ABEBA™ 7131127/39
 View more versions of this product
Product Code. 17181525
  • 800.00 DKK / 1 pair
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Description
  • Lightweight, comfortable black occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Black microfiber
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Specifications
Black
39
Unisex
Water Resistant
Microfiber
Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Low-cut
O2 SRC ESD