Learn More
Thermo Scientific X50 CLEANUP KIT, 300MG PSA, 150MG GCB, 900MG MGSO4, 15ML TUBES
Brand:  Thermo Scientific S2-15-OTH-US-KIT
1972.90 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
X50 CLEANUP KIT, 300MG PSA, 150MG GCB, 900MG MGSO4, 15ML TUBES
Product Code. 17319852
  • 1985.00 DKK / Pack of 50
This item is not returnable. View return policy
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific X50 CLEANUP KIT, 300MG PSA, 150MG GCB, 900MG MGSO4, 15ML TUBES >