Learn More
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test
Rapidly detect Clostridium difficile toxin A and B from human fecal samples with a simple, three-step procedure and results in 20 minutes.
Brand:  Thermo Scientific™ R24650
3140.29 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13235629
  • 4010.00 DKK / 20 tests
Description
Show More
Specifications
Specifications
2°C to 30°C
Xpect C. difficile Toxin A/B Test
Test devices, conjugate reagent 1 (3.6mL), conjugate reagent 2 (3.0mL), specimen diluent, positive control (2.0mL), negative control (2.0mL), disposable transfer pipettes, dilution tubes, wooden applicator sticks
20 Tests/Kit
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number266463 Certificate TypeCertificate of Analysis Date07/05/2025 Catalog NumberR24650
Lot Number216292 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog NumberR24650
Lot Number187342 Certificate TypeCertificate of Analysis Date04/11/2024 Catalog NumberR24650
Lot Number182089 Certificate TypeCertificate of Analysis Date19/10/2024 Catalog NumberR24650
Lot Number181002 Certificate TypeCertificate of Analysis Date14/10/2024 Catalog NumberR24650
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test > Xpect C. difficile Toxin A/B Test, 20/pk