missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, ≥98.5% xylenes + ethylbenzene basis (ethylbenzene ≤25%), ACS Reagent, Honeywell™
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
CAS | 1330-20-7 |
Density | 0.86g/mL at 25°C |
Quantity | 4 L |
Explosion Limit | 0.07 |
Ignition Point | 867°F |
Packaging | Glass Bottle |
Linear Formula | C6H4(CH3)2 |
MDL Number | MFCD00077264 |
Vapor Pressure | 18mmHg (37.7°C) |
UN Number | UN1307 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction