Learn More
YK-4-279, MedChemExpress
MedChemExpress YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.
Brand: Medchem Express HY-14507
Description
YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.Specifications
YK-4-279 | |
Off-White | |
C17H13Cl2NO4 | |
DMSO : 25 mg/mL (68.27 mM; Need ultrasonic) | |
366.2 | |
99.61% | |
Solid |
1037184-44-3 | |
Research | |
100 mg | |
ClC1=C2C(C(O)(CC(C3=CC=C(OC)C=C3)=O)C(N2)=O)=C(Cl)C=C1 | |
366.2 | |
Research | |
Cancer-programmed cell death |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only