missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1. Inhibits Ewing's sarcoma family tumor (ESFT) cell growth; induces apoptosis.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 366.2 |
Percent Purity | >99% |
Chemical Name or Material | YK 4-279 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
YK 4-279, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction