missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YM 58483, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
YM 58483 is a blocker of store-operated Ca2+ entry (SOCE), which mediates the activation of non-excitable cells (e.g. lymphocytes). Inhibits calcium release-activated calcium (CRAC) channels; suppresses thapsigargin -induced sustained Ca2+ influx (IC50 = 100 nM). Displays immuno-modulatory and anti-inflammatory effects; suppresses cytokine production and proliferation of T cells in vitro.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 421.32 |
Percent Purity | >99% |
Chemical Name or Material | YM 58483 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'