Learn More
Thermo Scientific Chemicals YP Base Medium
Brand: Thermo Scientific Chemicals H26407.36
Description
For maintaining and developing yeast in molecular microbiology procedures
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Powder | |
Plastic Bottle | |
500 g | |
Hygroscopic; Moisture sensitive |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10204717 | Certificate of Analysis | 04/08/2022 | H26407.36,H26407.22 |
10166054 | Certificate of Analysis | 23/11/2021 | H26407.36,H26407.22 |
10194873 | Certificate of Analysis | 23/11/2021 | H26407.36,H26407.22 |
I29W022 | Certificate of Analysis | 23/11/2021 | H26407.36,H26407.22 |
10230036 | Certificate of Analysis | 23/11/2021 | H26407.36,H26407.22 |