Learn More
Emma Safety Footwear Zandvoort XD Safety Shoes
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
1260.00 DKK
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15918751
|
Emma Safety Footwear
MM54886639 |
39 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15928751
|
Emma Safety Footwear
MM54886644 |
44 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15938751
|
Emma Safety Footwear
MM54886645 |
45 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15948751
|
Emma Safety Footwear
MM54886646 |
46 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15958751
|
Emma Safety Footwear
MM54886647 |
47 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15968751
|
Emma Safety Footwear
MM54886648 |
48 |
1260.00 DKK
1 pair |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Zandvoort XD | |
Oiled Nubuck |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Emma Safety Footwear Zandvoort XD Safety Shoes