Learn More
ZEOprep™ SH (Thiol) Functionalized
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. THIOL Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. The THIOL ligand makes an Ion Exchange material suitable for weak anion interactions.
1860.00 DKK - 36645.00 DKK
Specifications
Particle Size | 40 to 63 μm |
---|---|
Pore Size | 60 Å |
Technique | Chromatography |
Type | Silica Gel |
Material | Silica Gel |
Product Code | Brand | Description | Price | Quantity | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity | Availability | ||||
30049030
|
ZEOprep
300178 |
1 kg | 1 kg |
10760.00 DKK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30049022
|
ZEOprep
302095 |
5 kg | 5 kg |
36645.00 DKK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30048983
|
ZEOprep
300177 |
100 g | 100 g |
1860.00 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30049005
|
ZEOprep
302083 |
500 g | 500 g |
7000.00 DKK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- SH (Thiol) functionalized for Ion Exchange separations
- Idea for weak anion applications
- Irregular Amorphous Silica Gel
- High Purity
- European Manufacture
- Swiss accuracy and consistency
- Excellent batch to batch reproducability
- ISO Certification
- High Chemical and Mechanical Stability
- Narrow Particles Size Distribution
- 20 to 500 μm particles size range
- Available in 60 A or 90 A pore size
- High loadability
- Selection of pack sizes available
- Suitable for gravity, flash, prep and process scale
Specifications
40 to 63 μm | |
Chromatography | |
Silica Gel | |
Irregular |
60 Å | |
Silica Gel | |
SH (Thiol) Functionalized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
ZEOprep™ SH (Thiol) Functionalized