missing translation for 'onlineSavingsMsg'
Learn More
Zinc chloride, ACS, 97% min, Thermo Scientific Chemicals Product Code.: 11311669

Zinc chloride, ACS, 97% min, Thermo Scientific Chemicals

Product Code. 11311669
5 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11311669

Brand: Thermo Scientific Alfa Aesar 012307.A7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol

It is used as a catalyst, as a dehydrating and condensing agent, soldering flux, and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is likewise employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in organic synthesis, for example in the Friedel-Crafts acylatoin, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichlorideShow MoreShow Less
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Boiling Point 732°C
Melting Point 290°C
Physical Form Lump
Quantity 5 kg
Assay Percent Range 97%
Solubility Information Freely soluble in water,and dilute HCl. Soluble in alcohol,glycerol,ether and acetone.
Formula Weight 136.29
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.91
Chemical Name or Material Zinc chloride, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ21J026 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number012307.A7,012307.30,012307.A1
Lot NumberT25J040 Certificate TypeCertificate of Analysis Date14/08/2023 Catalog Number012307.A7,012307.30,012307.A1
Lot NumberS08I014 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog Number012307.A7,012307.30,012307.A1
Lot NumberQ28I034 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog Number012307.A7,012307.30,012307.A1
Lot NumberI14P34 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012307.A7,012307.30,012307.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc chloride, ACS, 97% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.