missing translation for 'onlineSavingsMsg'
Learn More

ZNF407 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP3-06058AF405

Product Code. 18310496

  • 3100.35 DKK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ZNF407 Polyclonal specifically detects ZNF407 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ZNF407
Polyclonal
Immunocytochemistry/Immunofluorescence
FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407
Produced in rabbits immunized with E. coli-derived Human ZNF407 fragment.
0.1 mL
Primary
Human
Purified
Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
55628
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers