missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF625 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567321
This item is not returnable.
View return policy
Description
Immunogen sequence: HTGEKPCSSNT SKGQGEKIA Highest antigen sequence identity to the following orthologs - mouse 55%, rat 55%.
ZNF625 may be involved in transcriptional regulation.Specifications
ZNF625 | |
Polyclonal | |
Unconjugated | |
ZNF625 | |
Zinc finger protein 625; ZNF625 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96I27 | |
ZNF625 | |
Recombinant Human ZNF625 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only