accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103745-39-7

CAS RN 103745-39-7

IUPAC Name: 5-(1,4-diazepane-1-sulfonyl)isoquinoline
Synonyms: fasudil 法舒地尔 fasudilum
Molecular Weight (g/mol): 291.37
Molecular Formula: C14H17N3O2S
InChi Key: NGOGFTYYXHNFQH-UHFFFAOYSA-N
SMILES: O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.