missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyn-3-ol, 97%, Thermo Scientific™

406.40 DKK - 1338.00 DKK

Chemical Identifiers

CAS 105-31-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00014408
InChI Key LTFTWJYRQNTCHI-UHFFFAOYSA-N
Synonym 1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook reference
PubChem CID 7746
IUPAC Name hex-1-yn-3-ol
SMILES CCCC(C#C)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302487
SDS View Documents
Thermo Scientific Alfa Aesar
L11549.06
5 g N/A N/A N/A
11312487
SDS View Documents
Thermo Scientific Alfa Aesar
L11549.14
25 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H226
Toxic if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P262-P264b-P270-P280-P301+P310-P303+P361+P353-P310-P330-P363-P370+P378q-P501c

H226-H301-H310

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-286-7

missing translation for 'rtecsNumber' : MR0181000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Hexyn-3-ol, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.