Learn More
BINDER™ Temperature and Humidity Calibration Certificate (1 Measuring Point)
For Use With: BINDER units series MKF, KMF, KBF, KBF P, KBF LQC, MKFT
Brand: BINDER™ DL30-0301
Description
- One humidity point of measurement
- Including certificate
- Humidity measurement point in center of chamber
Specifications
BINDER Chambers Series MKF, KMF, KBF, KBF P, KBF LQC, MKFT | |
Temperature and Humidity: 1 measuring point | |
Temperature and Humidity Measurement |
Calibration temperature and humidity - including certificate, one measuring point in center of chamber at 25°C / 60 % RH or at specified values | |
Calibration Certificate |