Learn More
Microspec™ 1 Roll Autoclave Tape
Autoclavable tape roll
Brand:  Microspec™ X2
 View more versions of this product
Product Code. 16695482
  • 43.30 DKK / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Description
  • Autoclavable
  • Different sizes available
Specifications
Specifications
Autoclavable
121°C
Non-sterile
25 mm x 50 m
1
Product Title
Microspec™ 1 Roll Autoclave Tape > 25 mm x 52 m